Skip to Content
Merck
All Photos(1)

Documents

477559

Sigma-Aldrich

4-Bromo-2-fluorobenzyl bromide

98%

Synonym(s):

4,α-Dibromo-2-fluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(F)CH2Br
CAS Number:
Molecular Weight:
267.92
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

33-36 °C (lit.)

SMILES string

Fc1cc(Br)ccc1CBr

InChI

1S/C7H5Br2F/c8-4-5-1-2-6(9)3-7(5)10/h1-3H,4H2

InChI key

XMHNLZXYPAULDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Bromo-2-fluorobenzyl bromide is a substituted benzyl bromide. Its density is 1.9094g/ml at 25°C.

Application

4-Bromo-2-fluorobenzyl bromide may be used in the synthesis of 1-(2-fluoro-4-(1-methyl-1H-pyrazol-4-yl)benzyl)indoline-2,3-dione.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas M Bridges et al.
Probe Reports from the NIH Molecular Libraries Program, 2010 Mar 31 (Updated 2010 Oct 29) (2011-03-25)
The specific aim of this project is to identify, via a functional high-throughput screening (HTS) approach, small molecule positive allosteric modulators (PAMs) and/or allosteric agonists of the M1 muscarinic acetylcholine receptor (mAChR) that are cell permeable, possess submicromolar potency, and
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 285-285 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service