Skip to Content
Merck
All Photos(1)

Documents

47346

Sigma-Aldrich

Fmoc-N-Me-D-Ala-OH

≥98.0% (TLC)

Synonym(s):

Fmoc-N-methyl-D-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19NO4
CAS Number:
Molecular Weight:
325.36
Beilstein:
6936712
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98.0% (TLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

C[C@@H](N(C)C(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C19H19NO4/c1-12(18(21)22)20(2)19(23)24-11-17-15-9-5-3-7-13(15)14-8-4-6-10-16(14)17/h3-10,12,17H,11H2,1-2H3,(H,21,22)/t12-/m1/s1

InChI key

JOFHWKQIQLPZTC-GFCCVEGCSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service