Skip to Content
Merck
All Photos(1)

Key Documents

461040

Sigma-Aldrich

2,4-Diphenyl-4-methyl-1-pentene

97%

Synonym(s):

α-Methylstyrene dimer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(CH3)2CH2C(C6H5)=CH2
CAS Number:
Molecular Weight:
236.35
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.569 (lit.)

bp

161 °C/5 mmHg (lit.)

density

0.99 g/mL at 25 °C (lit.)

SMILES string

CC(C)(CC(=C)c1ccccc1)c2ccccc2

InChI

1S/C18H20/c1-15(16-10-6-4-7-11-16)14-18(2,3)17-12-8-5-9-13-17/h4-13H,1,14H2,2-3H3

InChI key

ZOKCNEIWFQCSCM-UHFFFAOYSA-N

Application

Chain transfer agent.
CTA for styrene, ABS, SBR, and acrylics.

Features and Benefits

Odorless, does not cause discoloration.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Liver

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

293.4 °F - closed cup

Flash Point(C)

145.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service