Skip to Content
Merck
All Photos(1)

Key Documents

460451

Sigma-Aldrich

2-Hydroxyethyl stearate

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CO2CH2CH2OH
CAS Number:
Molecular Weight:
328.53
EC Number:
MDL number:
UNSPSC Code:
12162002

grade

technical grade

bp

149 °C (lit.)

mp

55-57 °C (lit.)

solubility

hexane and water: insoluble
toluene, ethanol and acetone: soluble

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCCO

Application

Plasticizer

Storage Class Code

11 - Combustible Solids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service