Skip to Content
Merck
All Photos(1)

Key Documents

453439

Sigma-Aldrich

Triisopropyl orthoformate

anhydrous, 99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH[OCH(CH3)2]3
CAS Number:
Molecular Weight:
190.28
Beilstein:
1700668
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

grade

anhydrous

Assay

99.5%

impurities

<0.005% water

evapn. residue

<0.0005%

refractive index

n20/D 1.397 (lit.)

bp

65-66 °C/18 mmHg (lit.)

density

0.854 g/mL at 25 °C (lit.)

SMILES string

CC(C)OC(OC(C)C)OC(C)C

InChI

1S/C10H22O3/c1-7(2)11-10(12-8(3)4)13-9(5)6/h7-10H,1-6H3

InChI key

FPIVAWNGRDHRSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service