Skip to Content
Merck
All Photos(1)

Key Documents

448125

Sigma-Aldrich

Piperazine-2,2,3,3,5,5,6,6-d8 dihydrochloride

≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
£87.45

£87.45

List Price£106.00Save 17%

Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 G
£87.45

About This Item

Empirical Formula (Hill Notation):
C4D8H2N2·2HCl
CAS Number:
Molecular Weight:
167.11
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

£87.45

List Price£106.00Save 17%

Please contact Customer Service for Availability

Request more information

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

form

solid

mp

>300 °C (lit.)

mass shift

M+8

SMILES string

Cl.Cl.[2H]C1([2H])NC([2H])([2H])C([2H])([2H])NC1([2H])[2H]

InChI

1S/C4H10N2.2ClH/c1-2-6-4-3-5-1;;/h5-6H,1-4H2;2*1H/i1D2,2D2,3D2,4D2;;

InChI key

CVVIJWRCGSYCMB-VHGLFXLXSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josef Dib et al.
Journal of mass spectrometry : JMS, 50(2), 407-417 (2015-03-25)
AdipoR agonists are small, orally active molecules capable of mimicking the protein adiponectin, which represents an adipokine with antidiabetic and antiatherogenic effects. Two adiponectin receptors were reported in the literature referred to as adipoR1 and adipoR2. Activation of these receptors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service