440760
Fluorotriphenylsilane
97%
Synonym(s):
Triphenylfluorosilane, Triphenylsilyl fluoride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C6H5)3SiF
CAS Number:
Molecular Weight:
278.40
Beilstein:
2941368
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
Recommended Products
Assay
97%
mp
62-64 °C (lit.)
SMILES string
F[Si](c1ccccc1)(c2ccccc2)c3ccccc3
InChI
1S/C18H15FSi/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H
InChI key
UBGXLEFOIVWVRP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Xiaogang Guo et al.
Materials (Basel, Switzerland), 12(2) (2019-01-16)
In this study, a novel superhydrophobic nano-aluminum/iron (III) oxide composite has been prepared by a facile one-step process of electrophoretic deposition, with wide potential applications. The optimal suspension included ethanol, acetyl-acetone, and the additives of fluorotriphenylsilane and perfluorodecyltriethoxysilane. The microstructure
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service