Skip to Content
Merck
All Photos(1)

Key Documents

438448

Sigma-Aldrich

Silver carbonate

99.999% trace metals basis

Synonym(s):

Disilver carbonate, Silver(I) carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag2CO3
CAS Number:
Molecular Weight:
275.75
Beilstein:
6936654
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: silver

mp

210 °C (dec.) (lit.)

density

6.08 g/mL at 25 °C (lit.)

SMILES string

[Ag]OC(=O)O[Ag]

InChI

1S/CH2O3.2Ag/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

KQTXIZHBFFWWFW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Silver carbonate is used in Koenigs-Knorr glycosylation. The product is used as a protective agent to prevent the sedimentation of silver nanoparticles (AgNPs).

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photocatalytic behavior and photo-corrosion of visible-light-active silver carbonate/titanium dioxide.
Wang Y, et al.
Materials Letters, 115, 85-88 (2014)
Silver carbonate and stability in colloidal silver: a by-product of the electric spark discharge method.
Tseng KH, et al.
J. Alloy Compounds, 493(1), 438-440 (2010)
A novel high-efficiency visible-light sensitive Ag 2 CO 3 photocatalyst with universal photodegradation performances: Simple synthesis, reaction mechanism and first-principles study.
Dong H, et al.
Applied Catalysis. B, Environmental, 134, 46-54 (2013)
Photocatalytic behavior and photo-corrosion of visible-light-active silver carbonate/titanium dioxide.
Wang Y, et al
Materials Letters, 115, 85-88 (2014)
Sukalyan Bhadra et al.
Journal of the American Chemical Society, 134(24), 9938-9941 (2012-06-12)
Decarboxylative Chan-Evans-Lam-type couplings are presented as a new strategy for the regiospecific construction of diaryl and alkyl aryl ethers starting from easily available aromatic carboxylic acids. They allow converting various aromatic carboxylate salts into the corresponding aryl ethers by reaction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service