Skip to Content
Merck
All Photos(1)

Documents

427799

Sigma-Aldrich

Polyethylene

average Mw ~35,000 by GPC, average Mn ~7,700 by GPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(CH2CH2)nH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

melt index

2,250 g/10 min (190 °C/2.16kg)

mol wt

average Mn ~7,700 by GPC
average Mw ~35,000 by GPC

viscosity

78 poise(150 °C, Brookfield Thermosel)(lit.)

mp

90 °C

acid number

<0.05 mg KOH/g

transition temp

softening point 104 °C (ring and ball)

density

0.906 g/mL at 25 °C

application(s)

battery manufacturing

InChI

1S/C2H4/c1-2/h1-2H2

InChI key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hot melt coating for paper, additive in cast moldings, candles, oil-based inks and hot melt adhesives.

Features and Benefits

Contributes high gloss, low moisture transfer, grease resistance and heat sealability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharine S Woods et al.
American journal of veterinary research, 74(4), 523-529 (2013-03-28)
To compare the diagnostic quality of bronchoalveolar lavage (BAL) fluid acquired from healthy dogs by manual aspiration via polyethylene tubing (MAPT) and via suction pump aspiration (SPA) with a suction trap connection. 12 healthy adult Beagles. BAL was performed with
[Technologies for manufacturing ultra-high molecular polyethylene-based porous structures for bone implants].
A V Maksimkin et al.
Meditsinskaia tekhnika, (2)(2), 16-20 (2013-06-26)
Łukasz Krakowxzyk et al.
Medical science monitor basic research, 19, 210-213 (2013-08-22)
The search for new surgical flap techniques and modifications of already existing ones is gaining increasing popularity. Progress in flap designing and harvesting have improved the functional and aesthetic results, especially in head and neck reconstruction. Ten pigs were used
Matteo Gumiero et al.
Food chemistry, 138(2-3), 1633-1640 (2013-02-16)
A high density polyethylene (HDPE)/calcium carbonate (CaCO(3)) film containing TiO(2) was prepared via blown film extrusion process. The photocatalytic properties of this film were evaluated by voltammetric, UV-Vis spectrophotometric and gas chromatographic measurements following the decomposition rate of suitably selected
Melody Yee-Man Wong et al.
Rapid communications in mass spectrometry : RCM, 27(6), 713-721 (2013-02-19)
Despite various porous materials having been widely adopted as spraying tips for direct sample analysis using electrospray ionization mass spectrometry (ESI-MS), the effect of surface property and porosity of spraying tip materials on their analytical performances is not clear. Investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service