Skip to Content
Merck
All Photos(1)

Documents

425273

Sigma-Aldrich

2,11,20,29-Tetra-tert-butyl-2,3-naphthalocyanine

Dye content 97 %

Synonym(s):

2,11,20,29-Tetra-tert -butyl-2,3-naphthophthalocyanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H58N8
CAS Number:
Molecular Weight:
939.20
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

composition

Dye content, 97%

λmax

784 nm

SMILES string

CC(C)(C)c1ccc2cc3c4nc(nc5[nH]c(nc6nc(nc7[nH]c(n4)c8cc9cc(ccc9cc78)C(C)(C)C)c%10cc%11cc(ccc%11cc6%10)C(C)(C)C)c%12cc%13cc(ccc%13cc5%12)C(C)(C)C)c3cc2c1

InChI

1S/C64H58N8/c1-61(2,3)41-17-13-33-25-45-49(29-37(33)21-41)57-65-53(45)70-58-51-31-39-23-43(63(7,8)9)19-15-35(39)27-47(51)55(67-58)72-60-52-32-40-24-44(64(10,11)12)20-16-36(40)28-48(52)56(68-60)71-59-50-30-38-22-42(62(4,5)6)18-14-34(38)26-46(50)54(66-59)69-57/h13-32H,1-12H3,(H2,65,66,67,68,69,70,71,72)

InChI key

FZJGGSUDHJDHRJ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service