Skip to Content
Merck
All Photos(1)

Key Documents

423319

Sigma-Aldrich

Dicyclohexyliodoborane solution

0.5 M in hexanes

Synonym(s):

Dicyclohexylboryl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2BI
CAS Number:
Molecular Weight:
304.02
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

form

liquid

reaction suitability

reagent type: reductant

concentration

0.5 M in hexanes

bp

68-69 °C

density

0.754 g/mL at 25 °C

SMILES string

IB(C1CCCCC1)C2CCCCC2

InChI

1S/C12H22BI/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-12H,1-10H2

InChI key

RWFGGTOYIFQXAO-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of vinyloxyboranes

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service