Skip to Content
Merck
All Photos(1)

Documents

420085

Sigma-Aldrich

(R)-2-Hydroxy-4-phenylbutyric acid

99% (sum of enantiomers)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2CH(OH)CO2H
CAS Number:
Molecular Weight:
180.20
Beilstein:
3201296
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Assay

99% (sum of enantiomers)

optical activity

[α]20/D −9.5°, c = 2.8 in ethanol

mp

114-117 °C (lit.)

SMILES string

O[C@H](CCc1ccccc1)C(O)=O

InChI

1S/C10H12O3/c11-9(10(12)13)7-6-8-4-2-1-3-5-8/h1-5,9,11H,6-7H2,(H,12,13)/t9-/m1/s1

InChI key

JNJCEALGCZSIGB-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service