Skip to Content
Merck
All Photos(1)

Documents

406406

Sigma-Aldrich

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzyl bromide

98%

Synonym(s):

1-(Bromomethyl)-2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)-2,3,5,6-tetrafluorobenzyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6F4CH2Br
CAS Number:
Molecular Weight:
310.99
Beilstein:
8423469
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.38 psi ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.448 (lit.)

density

1.864 g/mL at 25 °C (lit.)

SMILES string

Fc1c(F)c(c(F)c(F)c1CBr)C(F)(F)F

InChI

1S/C8H2BrF7/c9-1-2-4(10)6(12)3(8(14,15)16)7(13)5(2)11/h1H2

InChI key

WKPYRDRWTNJBQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzyl bromide (4-(trifluoromethyl)-2,3,5,6-tetrafluorobenzyl bromide) has been reported to be prepared in one step from α,α,α−2,3,5,6-heptafluoro-p-xylene.

Application

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzyl bromide (4-(trifluoromethyl)-2,3,5,6-tetrafluorobenzyl bromide, TTBB) may be employed as an alternate of pentafluorobenzyl bromide (PFBB) in electrophoric derivatization reactions prior to detection during gas chromatography-electron-capture negative ion mass spectrometry (GC-ECNI-MS). TTBB may be employed as derivatization reagent for the characterization and quantitation of DNA and hemoglobin adducts.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Saha et al.
Journal of chromatography, 641(2), 400-404 (1993-07-09)
4-(Trifluoromethyl)-2,3,5,6-tetrafluorobenzyl bromide (TTBB) was synthesized in a single step from alpha,alpha,alpha,2,3,5,6-heptafluoro-p-xylene. The purpose of TTBB is to function as an analogue of pentafluorobenzyl bromide (PFBB) in electrophoric derivatization reactions prior to detection by gas chromatography-electron-capture negative ion mass spectrometry (GC-ECNI-MS).
A Ranasinghe et al.
Chemical research in toxicology, 11(5), 520-526 (1998-06-20)
The analytical potential of gas chromatography/electron capture negative chemical ionization high-resolution mass spectrometry (HRMS) for characterization and quantitation of DNA and hemoglobin adducts was demonstrated using three model compounds: N2, 3-ethenoguanine (EG), 7-(2-hydroxyethyl)guanine (7-HEG), and N-(2-hydroxyethyl)valine (HEV). At a resolving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service