Skip to Content
Merck
All Photos(1)

Documents

405205

Sigma-Aldrich

Tetraphenylantimony(V) bromide

97%

Synonym(s):

NSC 129511, Tetraphenylstibonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)4SbBr
CAS Number:
Molecular Weight:
510.08
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

reaction suitability

core: antimony
reagent type: catalyst

mp

204 °C (dec.) (lit.)

SMILES string

Br[Sb](c1ccccc1)(c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/4C6H5.BrH.Sb/c4*1-2-4-6-5-3-1;;/h4*1-5H;1H;/q;;;;;+1/p-1

InChI key

MRSXZVCXTGNNFI-UHFFFAOYSA-M

Application

Catalyst for:
  • Chemo- and stereoselective addition of tin enolate to alpha-halo ketone
  • Selective reagent for synthesis of silyl enol ethers from cyclic ketones
  • Preparation of cyclic carbonates

Reactant for synthesis of tetraphenylantimony complexes of pyridine N-oxides

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service