Skip to Content
Merck
All Photos(1)

Key Documents

402141

Sigma-Aldrich

3-Formyl-6-isopropylchromone

99%

Synonym(s):

6-Isopropyl-4-oxo-4H-1-benzopyran-3-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12O3
CAS Number:
Molecular Weight:
216.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

99%

mp

98-100 °C (lit.)

SMILES string

CC(C)c1ccc2OC=C(C=O)C(=O)c2c1

InChI

1S/C13H12O3/c1-8(2)9-3-4-12-11(5-9)13(15)10(6-14)7-16-12/h3-8H,1-2H3

InChI key

FRRYMYQANNFABF-UHFFFAOYSA-N

Gene Information

human ... PTPN1(5770)

General description

3-Formyl-6-isopropylchromone (6-Isopropyl-4-oxo-4H-1-benzopyran-3-carboxaldehyde, 6-Isopropyl-4-oxo-4H-chromene-3-carbaldehyde), a pyrazine (paradiazine) derivative, is a heterocyclic building block. It is a heterocyclic six-membered aromatic compound bearing nitrogen atoms at para positions.
Combustion calorimetric experiments have been conducted to evaluate the enthalpy of combustion of 3-formyl-6-isopropylchromone.[1] Cytotoxic activity of 3-formyl-6-isopropylchromone against normal and tumor cells in vivo has been tested.[2]

Application

3-Formyl-6-isopropylchromone is the suitable reagent used in a study to investigate the multidrug resistance reversal by some 3- formylchromones in human colon cancer and mouse lymphoma cells transfected with the human MDR1 gene.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masami Kawase et al.
In vivo (Athens, Greece), 21(5), 829-834 (2007-11-21)
Several 3-formylchromone derivatives were examined for their tumor cell-cytotoxic, anti-Helicobacter pylori, urease inhibitory and anti-HIV activity. Comparing their relative cytotoxicity against four human tumor cell lines and three normal human cells, tumor cell-specific cytotoxicity was detected in some 3-formylchromone derivatives.
Enthalpies of combustion and formation of 3-formylchromones.
Flores H, et al.
Thermochimica Acta, 450(1), 35-37 (2006)
Zoltán Baráth et al.
In vivo (Athens, Greece), 20(5), 645-649 (2006-11-10)
Several new 3-formylchromone derivatives proved to be modifiers of multidrug resistance in mouse lymphoma cells and in human Colo320 colon cancer cells. There is apparently a structure-activity relationship between the antiproliferative multidrug resistance-reversing effect and the chemical structure of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service