Skip to Content
Merck
All Photos(1)

Key Documents

40205

Sigma-Aldrich

Ammonium hydroxide solution

≥25% NH3 in H2O, semiconductor grade PURANAL (Honeywell 17814)

Synonym(s):

Ammonia aqueous, Ammonia water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein:
3587154
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

semiconductor grade PURANAL (Honeywell 17814)

CofA

specification on request

expl. lim.

27 %

concentration

≥25% NH3 in H2O

density

0.9 g/mL at 25 °C (lit.)

SMILES string

[NH4+].[OH-]

InChI

1S/H3N.H2O/h1H3;1H2

InChI key

VHUUQVKOLVNVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium hydroxide is a weak base with an increasing dissociation upon dilution. The base serves as a;
  • precipitant for the R2O3 group of elements,
  • complexing agent,
  • dissolution medium.7

Application

Ammonium hydroxide may be used;
  • to clean wafers.1
  • to prepare magnetic iron oxide nanoparticles[1]
  • in the recovery of cellulose from sugarcane bagasse. The process enhances hydrolysis and -production of bioethanol[2]
  • in bi-directional perfusion technique of human periodontal ligament cell sheets[3]
  • as a mobile phase in the liquid chromatography for prebiotic galactooligosaccharides[4]

Legal Information

PURANAL is a trademark of Honeywell Specialty Chemicals Seelze GmbH

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A systematic study of transfection efficiency and cytotoxicity in HeLa cells using iron oxide nanoparticles prepared with organic and inorganic bases
Calmon MF, et al.
Colloids and Surfaces. B, Biointerfaces, 100(1), 177-184 (2012)
Zhisheng Zhu et al.
Bioresource technology, 119, 199-207 (2012-06-26)
An efficient pretreatment method using NH(4)OH-H(2)O(2) and ionic liquid (IL) was developed for the recovery of cellulose from sugarcane bagasse (SCB). The regenerated SCB from the combined pretreatment exhibited significantly enhanced enzymatic digestibility with an efficiency of 91.4% after 12h
O Hernández-Hernández et al.
Journal of chromatography. A, 1220, 57-67 (2011-12-23)
Three different stationary phases (sulfoalkylbetaine zwitterionic, polyhydroxyethyl aspartamide and ethylene bridge hybrid (BEH) with trifunctionally bonded amide), operating at hydrophilic interaction liquid chromatographic (HILIC) mode, have been assayed and compared for the analysis of complex mixtures of galactooligosaccharides (GOS). Chromatographic
A Farag et al.
Journal of dental research, 93(12), 1313-1319 (2014-10-02)
The periodontal ligament is the key tissue facilitating periodontal regeneration. This study aimed to fabricate decellularized human periodontal ligament cell sheets for subsequent periodontal tissue engineering applications. The decellularization protocol involved the transfer of intact human periodontal ligament cell sheets
Konstantin Chingin et al.
Analytical chemistry, 84(15), 6856-6862 (2012-07-12)
We introduce an online multiple-junction capillary isoelectric focusing fractionator (OMJ-CIEF) for separation of biological molecules in solution by pI. In OMJ-CIEF, the separation capillary is divided into seven equal sections joined with each other via tubular Nafion membrane insertions. Each

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service