Skip to Content
Merck
All Photos(1)

Documents

392065

Sigma-Aldrich

2-(Allylthio)benzimidazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2S
CAS Number:
Molecular Weight:
190.26
UNSPSC Code:
12352200

Assay

97%

mp

139-141 °C (lit.)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service