384380
[3-(Heptafluoroisopropoxy)propyl]trichlorosilane
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Assay
97%
refractive index
n20/D 1.371 (lit.)
bp
39 °C/0.5 mmHg (lit.)
density
1.497 g/mL at 25 °C (lit.)
SMILES string
FC(F)(F)C(F)(OCCC[Si](Cl)(Cl)Cl)C(F)(F)F
Packaging
Packaged in glass bottles
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service