Recommended Products
Assay
≥98.0% (NT)
optical activity
[α]20/D +1.2±0.4°, c = 5% in 1 M HCl
mp
199-201 °C (dec.)
200 °C (dec.) (lit.)
storage temp.
2-8°C
SMILES string
N[C@@H](Cc1cc(I)c(O)c(I)c1)C(O)=O
InChI
1S/C9H9I2NO3/c10-5-1-4(2-6(11)8(5)13)3-7(12)9(14)15/h1-2,7,13H,3,12H2,(H,14,15)/t7-/m0/s1
InChI key
NYPYHUZRZVSYKL-ZETCQYMHSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
Intermediate in the biosynthesis and alternative pathways of metabolism of thyroid hormones. Substrate for the assay of halogenated tyrosine and thyroid hormone aminotransferase
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Purification and properties of halogenated tyrosine and thyroid hormone transaminase from rat kidney mitochondria.
The Journal of biological chemistry, 242(1), 73-81 (1967-01-10)
The Journal of clinical investigation, 72(4), 1234-1245 (1983-10-01)
Studies were performed to elucidate the nature of the pathway of hepatic thyroxine (T4) metabolism that is activated by inhibitors of liver catalase. For this purpose, the metabolism of T4 in homogenates of rat liver was monitored with T4 labeled
Methods in Enzymology, 17A, 660-660 (1970)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service