Skip to Content
Merck
All Photos(1)

Key Documents

379425

Sigma-Aldrich

Yttrium isopropoxide oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OY5(OCH(CH3)2)13
CAS Number:
Molecular Weight:
1228.66
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

composition

Y, 35-37% complexometric

reaction suitability

core: yttrium
reagent type: catalyst

SMILES string

CC(C)O[Y]=O.CC(C)O[Y](OC(C)C)OC(C)C.CC(C)O[Y](OC(C)C)OC(C)C.CC(C)O[Y](OC(C)C)OC(C)C.CC(C)O[Y](OC(C)C)OC(C)C

InChI

1S/13C3H7O.O.5Y/c13*1-3(2)4;;;;;;/h13*3H,1-2H3;;;;;;/q13*-1;;+1;4*+3

InChI key

QUIINUCCTZSPAV-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service