Skip to Content
Merck
All Photos(1)

Key Documents

374490

Sigma-Aldrich

Benzyltrichlorosilane

95%

Synonym(s):

Trichloro-p-tolylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SiCl3
CAS Number:
Molecular Weight:
225.57
Beilstein:
2935870
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

95%

refractive index

n20/D 1.524 (lit.)

bp

218-220 °C (lit.)

density

1.273 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(cc1)[Si](Cl)(Cl)Cl

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service