Skip to Content
Merck
All Photos(1)

Key Documents

369063

Sigma-Aldrich

2-Bromo-4-chloro-6-fluoroaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(Cl)(F)NH2
CAS Number:
Molecular Weight:
224.46
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

41-43 °C (lit.)

functional group

bromo
chloro
fluoro

SMILES string

Nc1c(F)cc(Cl)cc1Br

InChI

1S/C6H4BrClFN/c7-4-1-3(8)2-5(9)6(4)10/h1-2H,10H2

InChI key

HBHBARSMRVAINH-UHFFFAOYSA-N

General description

2-Bromo-4-chloro-6-fluoroaniline is an aniline derivative.

Application

2-Bromo-4-chloro-6-fluoroaniline may be used in the preparation of fragment A of an orally bioavailable and CNS penetrant bradykinin1antagonist.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Process development of a potent bradykinin 1 antagonist.
Menzel K, et al.
Organic Process Research & Development, 13(3), 519-524 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service