Skip to Content
Merck
All Photos(1)

Key Documents

366250

Sigma-Aldrich

Pyridinium chlorochromate on alumina

approx. 20 wt. % on basic alumina

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: alcohol reactive

SMILES string

O[Cr](Cl)(=O)=O.c1ccncc1

InChI

1S/C5H5N.ClH.Cr.H2O.2O/c1-2-4-6-5-3-1;;;;;/h1-5H;1H;;1H2;;/q;;+2;;;/p-2

InChI key

JWIPGAFCGUDKEY-UHFFFAOYSA-L

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Ox. Sol. 2 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service