Skip to Content
Merck
All Photos(1)

Key Documents

36380

Sigma-Aldrich

Methyl 2,3-dichloropropionate

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CHClCOOCH3
CAS Number:
Molecular Weight:
157.00
Beilstein:
1701891
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.453 (lit.)
n20/D 1.453

bp

52-55 °C/2 mmHg (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(Cl)CCl

InChI

1S/C4H6Cl2O2/c1-8-4(7)3(6)2-5/h3H,2H2,1H3

InChI key

OFHMODDLBXETIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service