Skip to Content
Merck
All Photos(1)

Documents

363545

Sigma-Aldrich

2-(Methylsulfonyl)ethanol

98%

Synonym(s):

2-Hydroxyethyl methyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO2CH2CH2OH
CAS Number:
Molecular Weight:
124.16
Beilstein:
1746915
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

bp

148-149 °C (lit.)

mp

31-33 °C (lit.)

SMILES string

CS(=O)(=O)CCO

InChI

1S/C3H8O3S/c1-7(5,6)3-2-4/h4H,2-3H2,1H3

InChI key

KFTYFTKODBWKOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reaction of 2-(methylsulfonyl)ethanol with cotton cellulose yields methylsulfonylethyl substituents in the 2-O- to 6-O-positions of the monosubstituted D-glucopyranosyl unit of cotton cellulose.

Application

2-(Methylsulfonyl)ethanol may be used as reagent in the synthesis of phenols from aryl fluorides. It may be used in the preparation of 2-(methylsulfonyl)ethyl N,N-bis(2-chloroethyl)phosphorodiamidate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monish Jain et al.
Journal of medicinal chemistry, 47(15), 3843-3852 (2004-07-09)
A series of sulfonyl-group containing analogues of aldophosphamide (Aldo) were synthesized as potential anticancer prodrugs that liberate the cytotoxic phosphoramide mustards (PM, IPM, and tetrakis-PM) via beta-elimination, a nonenzymatic activation mechanism. Kinetic studies demonstrated that all these compounds spontaneously liberate
Reagent effects on distribution of methylsulfonylethyl substituents in the D-glucopyranosyl unit of cotton cellulose.
Rowland SP, et al.
Canadian Journal of Chemistry, 46(3), 451-457 (1968)
Sebastian Bonardd et al.
Polymers, 13(3) (2021-01-28)
In this work, polymethacrylates containing sulfonyl and nitrile functional groups were successfully prepared by conventional radical polymerization and reversible addition-fragmentation chain-transfer polymerization (RAFT). The thermal and dielectric properties were evaluated, for the first time, considering differences in their molecular weights
Mild conversion of electron deficient aryl fluorides to phenols using 2-(methylsulfonyl) ethanol.
Rogers JF and Green DF.
Tetrahedron Letters, 43(19), 3585-3587 (2002)
S G Cohen et al.
Journal of medicinal chemistry, 28(9), 1309-1313 (1985-09-01)
Reversible inhibitors for acetylcholinesterase, AcChE, have been studied. Sterically similar alcohols with tetra-substituted uncharged beta groups, (CH3)3SiCH2CH2OH (I), (CH3)3CCH2CH2OH (IA), and CH3S(O2)CH2CH2OH (VII), bind similarly, KI = 3-9 mM, and each binds similarly to its acetate substrate; cationic analogues, (CH3)3N+CH2CH2OH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service