Skip to Content
Merck
All Photos(2)

Documents

362948

Sigma-Aldrich

Barium bis(2,2,6,6-tetramethyl-3,5-heptanedionate) hydrate

Synonym(s):

Ba(TMHD)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(OCC(CH3)3CHCOC(CH3)3)2 · xH2O
CAS Number:
Molecular Weight:
503.86 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Ba, ≥26.3% gravimetric (water corrected with sulfuric acid)
Degree of hydration, ~1

reaction suitability

core: barium

mp

175-180 °C (lit.)

SMILES string

[H]O[H].CC(C)(C)C(=O)\C=C(/O[Ba]O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/2C11H20O2.Ba.H2O/c2*1-10(2,3)8(12)7-9(13)11(4,5)6;;/h2*7,12H,1-6H3;;1H2/q;;+2;/p-2/b2*8-7-;;

InChI key

VCALGUJWYYNHDY-ZJCTYWPYSA-L

Application

Used in the preparation of superconducting thin films.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thin Solid Films, 182, 265-265 (1989)
Chem. Abstr., 112, 149887h-149887h (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service