Recommended Products
Assay
98%
mp
42-45 °C (lit.)
SMILES string
Cc1cc(c(Cl)cc1F)[N+]([O-])=O
InChI
1S/C7H5ClFNO2/c1-4-2-7(10(11)12)5(8)3-6(4)9/h2-3H,1H3
InChI key
SJDPAVRCQNFVDM-UHFFFAOYSA-N
Application
4-Chloro-2-fluoro-5-nitrotoluene may be used in the synthesis of two new C-11 labeled analogs of N,N-dimethyl-2-(2′-amino-4′-hydroxymethyl-phenylthio)benzylamine, which were further evaluated as serotonin transporter imaging agents.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nuclear medicine and biology, 37(5), 593-603 (2010-07-09)
Carbon-11 (C-11) N,N-dimethyl-2-(2'-amino-4'-hydroxymethyl-phenylthio)benzylamine ([(11)C]HOMADAM) has been reported as highly specific and selective positron emission tomography (PET) radiotracer showing fast kinetics for the human brain serotonin transporter (SERT). In our continued effort to develop appropriate PET SERT radioligand that can be
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service