Skip to Content
Merck
All Photos(2)

Key Documents

330264

Sigma-Aldrich

3,5-Dimethoxybenzyl chloride

99%

Synonym(s):

1-(Chloromethyl)-3,5-dimethoxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2Cl
CAS Number:
Molecular Weight:
186.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

115-118 °C/0.4 mmHg (lit.)

mp

46-48 °C (lit.)

functional group

chloro

SMILES string

COc1cc(CCl)cc(OC)c1

InChI

1S/C9H11ClO2/c1-11-8-3-7(6-10)4-9(5-8)12-2/h3-5H,6H2,1-2H3

InChI key

CCAWDIFJOBKBSE-UHFFFAOYSA-N

Application

3,5-Dimethoxybenzyl chloride was used in preparation of benzyl 7-(3,5-dimethoxyphenylacetoxy)octanoate, a precursor of curvularin.[1] It was also used in the preparation of diethyl (3,5-dimethoxybenzyl) phosphonate via Wittig-Horner reaction.[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of (?)-dimethyl curvularin based on the palladium-catalyzed carbonylation of 3, 5-dimethoxybenzyl chloride using a butadiene telomer as a building block.
Takahashi T, et al.
Tetrahedron Letters, 21(40), 3885-3888 (1980)
Guoliang Chen et al.
Chemical & pharmaceutical bulletin, 53(12), 1587-1590 (2005-12-06)
Seventeen novel resveratrol derivatives were synthesized. Their anti-inflammatory activities were tested on xylene-induced mouse ear edema. The pharmacological results showed that some compounds have potent anti-inflammatory activities.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service