Skip to Content
Merck
All Photos(1)

Documents

330248

Sigma-Aldrich

5-Chloro-1-ethyl-2-methylimidazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9ClN2
CAS Number:
Molecular Weight:
144.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.499 (lit.)

bp

101 °C/12 mmHg (lit.)

density

1.141 g/mL at 25 °C (lit.)

SMILES string

CCn1c(Cl)cnc1C

InChI

1S/C6H9ClN2/c1-3-9-5(2)8-4-6(9)7/h4H,3H2,1-2H3

InChI key

RHQYMLWGPGEEOU-UHFFFAOYSA-N

Application

5-Chloro-1-ethyl-2-methylimidazole was employed as additive to study the effect of additives on the performance of dye-sensitized TiO2 solar cells based on ionic liquid electrolyte.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficiency enhancement of dye-sensitized solar cells with addition of additives (single/binary) to ionic liquid electrolyte.
Lee HS, et al.
Bulletin of Materials Science, 35(6), 1003-1010 (2012)
Maria Duda et al.
Developmental cell, 48(2), 245-260 (2019-01-30)
As tissues develop, they are subjected to a variety of mechanical forces. Some of these forces are instrumental in the development of tissues, while others can result in tissue damage. Despite our extensive understanding of force-guided morphogenesis, we have only
Devon L Moose et al.
Cell reports, 30(11), 3864-3874 (2020-03-19)
During metastasis, cancer cells are exposed to potentially destructive hemodynamic forces including fluid shear stress (FSS) while en route to distant sites. However, prior work indicates that cancer cells are more resistant to brief pulses of high-level FSS in vitro relative to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service