About This Item
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.645 (lit.)
bp
168 °C/20 mmHg (lit.)
density
1.19 g/mL at 25 °C (lit.)
SMILES string
CCNc1ccccc1[N+]([O-])=O
InChI
1S/C8H10N2O2/c1-2-9-7-5-3-4-6-8(7)10(11)12/h3-6,9H,2H2,1H3
InChI key
CQIKVOWCSGXCCG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service