Skip to Content
Merck
All Photos(1)

Key Documents

305855

Sigma-Aldrich

(3-(Dimethylamino)propyl)triphenylphosphonium bromide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2CH2P(C6H5)3(Br)
CAS Number:
Molecular Weight:
428.34
Beilstein:
4637548
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

mp

193-196 °C

SMILES string

[Br-].CN(C)CCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C23H27NP.BrH/c1-24(2)19-12-20-25(21-13-6-3-7-14-21,22-15-8-4-9-16-22)23-17-10-5-11-18-23;/h3-11,13-18H,12,19-20H2,1-2H3;1H/q+1;/p-1

InChI key

SSWPSKSQQSJKKF-UHFFFAOYSA-M

Application

Reactant for preparation of:
  • Topoisomerase I-targeting antitumor agents
  • C15-C21 stereopentad of discodermolide via cleavage of aminoglycosides, double-bond isomerization, and selective hydroxyl group protection
  • Diphenyl amine based sodium channel blockers, effective against hNav1.2

  • Reactant for Wittig reaction with benzaldehyde

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service