About This Item
Recommended Products
Assay
97%
refractive index
n20/D 1.445 (lit.)
bp
137-140 °C (lit.)
density
0.815 g/mL at 25 °C (lit.)
functional group
amine
SMILES string
CN(C)\C=N\C(C)(C)C
InChI
1S/C7H16N2/c1-7(2,3)8-6-9(4)5/h6H,1-5H3/b8-6+
InChI key
PHNRCFCIKPZSFS-SOFGYWHQSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
93.2 °F - closed cup
Flash Point(C)
34 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service