Recommended Products
Assay
98%
mp
129-132 °C (lit.)
functional group
ester
SMILES string
CCOC(=O)c1c(C)cc(O)cc1O
InChI
1S/C10H12O4/c1-3-14-10(13)9-6(2)4-7(11)5-8(9)12/h4-5,11-12H,3H2,1-2H3
InChI key
UQSRXQMIXSZGLA-UHFFFAOYSA-N
General description
Ethyl 2,4-dihydroxy-6-methylbenzoate is also referred as ethyl orsellinate. It was isolated from chloroform extract of Peltigera aphthosa. It is one of the product formed during alcoholyses of lecanoric acid.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Antibacterial activity of orsellinates.
Brazilian Journal of Microbiology, 34(3), 194-196 (2003)
Antimicrobial agents and chemotherapy, 28(2), 289-292 (1985-08-01)
Antimicrobial screening of several lichen species and subsequent isolation and structure elucidation of active compounds revealed that the hydrolysis products of certain lichen metabolites, i.e., depsides, were active against gram-negative bacteria and fungi as well as gram-positive bacteria. The active
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service