Skip to Content
Merck
All Photos(1)

Key Documents

296627

Sigma-Aldrich

N,N-Diethylsalicylamide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO2
CAS Number:
Molecular Weight:
193.24
MDL number:
Pricing and availability is not currently available.

Assay

99%

mp

93-97 °C (lit.)

SMILES string

CCN(CC)C(=O)c1ccccc1O

InChI

1S/C11H15NO2/c1-3-12(4-2)11(14)9-7-5-6-8-10(9)13/h5-8,13H,3-4H2,1-2H3

InChI key

ZVYXEXAXXWINEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service