Skip to Content
Merck
All Photos(1)

Documents

288209

Sigma-Aldrich

Tetraphenylgermane

96%

Synonym(s):

Tetraphenyl germanium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ge(C6H5)4
CAS Number:
Molecular Weight:
381.06
EC Number:
MDL number:
UNSPSC Code:
12352300

Assay

96%

mp

230-235 °C (lit.)

SMILES string

c1ccc(cc1)[Ge](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C24H20Ge/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24/h1-20H

InChI key

ILEXMONMGUVLRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service