Skip to Content
Merck
All Photos(1)

Key Documents

278858

Sigma-Aldrich

3-Iodo-4-methylbenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H3(CH3)CO2H
CAS Number:
Molecular Weight:
262.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

210-212 °C (lit.)

SMILES string

Cc1ccc(cc1I)C(O)=O

InChI

1S/C8H7IO2/c1-5-2-3-6(8(10)11)4-7(5)9/h2-4H,1H3,(H,10,11)

InChI key

LDDHMKANNXWUAK-UHFFFAOYSA-N

Application

3-Iodo-4-methylbenzoic acid has been used in the preparation of:
  • unlabeled N-succinimidyl 4-guanidinomethyl-3-iodobenzoate (SGIMB)[1]
  • boc-protected derivative of SGIMB (Boc-SGMIB)[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Vaidyanathan et al.
Bioconjugate chemistry, 12(3), 428-438 (2001-05-17)
The objective of this study was to develop an acylation agent for the radioiodination of monoclonal antibodies that would maximize retention of the label in tumor cells following receptor- or antigen-mediated internalization. The strategy taken was to add a polar
Ganesan Vaidyanathan et al.
Nature protocols, 2(2), 282-286 (2007-04-05)
This protocol describes a detailed procedure for the synthesis of N-succinimidyl 4-guanidinomethyl-3-[*I]iodobenzoate ([*I]SGMIB), an agent useful in the radio-iodination of proteins, including monoclonal Abs, and peptides that undergo internalization after receptor or antigen binding. In this procedure, the tin precursor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service