Skip to Content
Merck
All Photos(2)

Key Documents

275298

Sigma-Aldrich

3,5-Dihydroxy-2-naphthoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C10H5CO2H
CAS Number:
Molecular Weight:
204.18
Beilstein:
3287771
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

275-280 °C (lit.)

SMILES string

OC(=O)c1cc2cccc(O)c2cc1O

InChI

1S/C11H8O4/c12-9-3-1-2-6-4-8(11(14)15)10(13)5-7(6)9/h1-5,12-13H,(H,14,15)

InChI key

YELLAPKUWRTITI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yinggang Luo et al.
The Journal of biological chemistry, 283(21), 14694-14702 (2008-04-05)
Neocarzinostatin, a clinical anticancer drug, is the archetypal member of the chromoprotein family of enediyne antitumor antibiotics that are composed of a nonprotein chromophore and an apoprotein. The neocarzinostatin chromophore consists of a nine-membered enediyne core, a deoxyaminosugar, and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service