271500
Lithium 3,5-diiodosalicylate
99%
Synonym(s):
2-Hydroxy-3,5-diiodobenzoic acid lithium salt, 3,5-Diiodo-2-hydroxybenzoic acid lithium salt, 3,5-Diiodosalicylic acid lithium salt, LIS
About This Item
Recommended Products
Assay
99%
SMILES string
[Li+].Oc1c(I)cc(I)cc1C([O-])=O
InChI
1S/C7H4I2O3.Li/c8-3-1-4(7(11)12)6(10)5(9)2-3;/h1-2,10H,(H,11,12);/q;+1/p-1
InChI key
HLBRJWWTLIAOTE-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
replaced by
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service