Skip to Content
Merck
All Photos(1)

Documents

270830

Sigma-Aldrich

1,1-Dichloro-3,3-dimethylbutane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2CHCl2
CAS Number:
Molecular Weight:
155.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.4388 (lit.)

bp

146-148 °C (lit.)

mp

−56 °C (lit.)

density

1.027 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)CC(Cl)Cl

InChI

1S/C6H12Cl2/c1-6(2,3)4-5(7)8/h5H,4H2,1-3H3

InChI key

BLRPQFOUUPCTED-UHFFFAOYSA-N

General description

The coupling of 1,1-dichloro-3,3-dimethylbutane and CD2Cl2 with nBuMgCl was studied.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zsolt Csok et al.
Journal of the American Chemical Society, 130(26), 8156-8157 (2008-06-06)
A new pincer-type bis(amino)amine (NN2) ligand and its lithium and nickel complexes, including Ni(II) methyl, ethyl, and phenyl complexes, were synthesized. The Ni(II) alkyl complexes react cleanly with alkyl halides including chlorides to form C-C coupled products and Ni(II) halides.
Sedat Dogan et al.
Nanoscale, 7(11), 4875-4883 (2015-02-13)
Solution-processable, two-dimensional semiconductors are promising optoelectronic materials which could find application in low-cost solar cells. Lead sulfide nanocrystals raised attention since the effective band gap can be adapted over a wide range by electronic confinement and observed multi-exciton generation promises

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service