Skip to Content
Merck
All Photos(1)

Key Documents

263796

Sigma-Aldrich

Vinyl chloroformate

94%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHOCOCl
CAS Number:
Molecular Weight:
106.51
Beilstein:
1741679
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

vapor pressure

0.88 psi ( 20 °C)

Assay

94%

contains

100 ppm BHT as inhibitor

refractive index

n20/D 1.41 (lit.)

bp

68-69 °C/735 mmHg (lit.)

density

1.166 g/mL at 20 °C (lit.)

SMILES string

ClC(=O)OC=C

InChI

1S/C3H3ClO2/c1-2-6-3(4)5/h2H,1H2

InChI key

PQLFROTZSIMBKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service