263486
2-(Trifluoromethyl)benzylamine
98%
Synonym(s):
2-(Aminomethyl)benzotrifluoride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CF3C6H4CH2NH2
CAS Number:
Molecular Weight:
175.15
Beilstein:
1103424
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
98%
refractive index
n20/D 1.471 (lit.)
density
1.249 g/mL at 25 °C (lit.)
SMILES string
NCc1ccccc1C(F)(F)F
InChI
1S/C8H8F3N/c9-8(10,11)7-4-2-1-3-6(7)5-12/h1-4H,5,12H2
InChI key
ZSKQIFWUTUZAGF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
2-(Trifluoromethyl)benzylamine may be used in chemical synthesis.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
156.2 °F - closed cup
Flash Point(C)
69 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service