Skip to Content
Merck
All Photos(2)

Key Documents

260053

Sigma-Aldrich

2-Bromo-6-nitrotoluene

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
£50.50

£50.50

List Price£101.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
£50.50

About This Item

Linear Formula:
CH3C6H3(NO2)Br
CAS Number:
Molecular Weight:
216.03
Beilstein:
2502581
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

£50.50

List Price£101.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

bp

143 °C/22 mmHg (lit.)

mp

38-40 °C (lit.)

functional group

bromo
nitro

SMILES string

Cc1c(Br)cccc1[N+]([O-])=O

InChI

1S/C7H6BrNO2/c1-5-6(8)3-2-4-7(5)9(10)11/h2-4H,1H3

InChI key

LYTNSGFSAXWBCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromo-6-nitrotoluene has been used:
  • as starting reagent in total synthesis of N-acetyl methyl ester of (±)-clavicipitic acids[1]
  • in synthesis of carbazomadurin A, highly oxygenated neuronal cell protecting carbazole alkaloid[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hans-Joachim Knölker et al.
Chemical communications (Cambridge, England), (10)(10), 1170-1171 (2003-06-05)
The highly oxygenated neuronal cell protecting carbazole alkaloid carbazomadurin A was synthesized in nine steps and 11% overall yield from isovanillic acid.
Palladium-catalyzed reactions in the synthesis of 3-and 4-substituted indoles. 2. Total synthesis of the N-acetyl methyl ester of (?)-clavicipitic acids.
Harrington PJ, et al.
Journal of the American Chemical Society, 109(14), 4335-4338 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service