Skip to Content
Merck
All Photos(1)

Key Documents

25619

Sigma-Aldrich

4-Chloro-3-nitro-5-sulfamoylbenzoic acid

technical, ≥90% (S)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5ClN2O6S
CAS Number:
Molecular Weight:
280.64
Beilstein:
820150
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (S)

mp

227-231 °C

SMILES string

NS(=O)(=O)c1cc(cc(c1Cl)[N+]([O-])=O)C(O)=O

InChI

1S/C7H5ClN2O6S/c8-6-4(10(13)14)1-3(7(11)12)2-5(6)17(9,15)16/h1-2H,(H,11,12)(H2,9,15,16)

InChI key

ACYLUAGCBGTEJF-UHFFFAOYSA-N

Application

4-Chloro-3-nitro-5-sulfamoylbenzoic acid has been used in preparation of 4-((2-(2-hydroxyphenyl)quinolin-3-yl)methylamino)-3-nitro-5-sulfamoylbenzoic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naveen Mulakayala et al.
Bioorganic & medicinal chemistry, 20(2), 759-768 (2011-12-29)
A facile and catalyst free synthesis of 6H-1-benzopyrano[4,3-b]quinolin-6-ones has been accomplished via the reaction of 4-chloro-2-oxo-2H-chromene-3-carbaldehyde with various aromatic amines in the presence of ultrasound. Some of these compounds were converted to the corresponding 2-(3-(hydroxymethyl)quinolin-2-yl)phenols and further structure elaboration of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service