Skip to Content
Merck
All Photos(1)

Key Documents

254967

Sigma-Aldrich

2-(Trifluoromethyl)benzyl bromide

96%

Synonym(s):

α′-Bromo-α,α,α-trifluoro-o-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CH2Br
CAS Number:
Molecular Weight:
239.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.494 (lit.)

bp

72 °C/7.5 mmHg (lit.)

density

1.571 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

FC(F)(F)c1ccccc1CBr

InChI

1S/C8H6BrF3/c9-5-6-3-1-2-4-7(6)8(10,11)12/h1-4H,5H2

InChI key

TXVVVEUSVBLDED-UHFFFAOYSA-N

Application

2-(Trifluoromethyl)benzyl bromide may be used in chemical synthesis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

William L Scott et al.
Journal of combinatorial chemistry, 11(1), 14-33 (2008-12-25)
Distributed Drug Discovery (D(3)) proposes solving large drug discovery problems by breaking them into smaller units for processing at multiple sites. A key component of the synthetic and computational stages of D(3) is the global rehearsal of prospective reagents and
William L Scott et al.
Journal of combinatorial chemistry, 11(1), 34-43 (2008-12-25)
For the successful implementation of Distributed Drug Discovery (D(3)) (outlined in the accompanying Perspective), students, in the course of their educational laboratories, must be able to reproducibly make new, high quality, molecules with potential for biological activity. This article reports

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service