Skip to Content
Merck
All Photos(1)

Key Documents

253561

Sigma-Aldrich

12-DOXYL-stearic acid, free radical

Synonym(s):

2-(10-Carboxydecyl)-2-hexyl-4,4-dimethyl-3-oxazolidinyloxy, free radical

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H43NO4
CAS Number:
Molecular Weight:
385.58
UNSPSC Code:
12352200

Application

Spin label for membrane fluidity studies.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Fukushima et al.
Toxicology and applied pharmacology, 102(2), 205-218 (1990-02-01)
The effects of phenylhydrazine (PHZ) and monomethylhydrazine (MMH) on the deformability of human erythrocytes and ghosts in relation to Heinz body formation are investigated using the sensitive flow EPR (electron paramagnetic resonance) method. The decrease in deformability starts even before
J M Burt
The American journal of physiology, 256(4 Pt 1), C913-C924 (1989-04-01)
The influence of doxyl stearic acids (DSAs) on gap junctional conductance (gj) between pairs of neonatal rat heart cells was studied. DSAs are spin probes that perturb the membrane at different depths depending on position of the doxyl group on
S M Musser et al.
Biophysical journal, 65(6), 2348-2359 (1993-12-01)
It has been known for some time that dicyclohexylcarbodiimide (DCCD) inhibits the proton translocation function of the cytochrome c oxidase complex (CcO) and that there is one major site in subunit III which is modified upon reaction with DCCD (Glu-90
M S Moxness et al.
Biochemistry, 35(22), 7181-7187 (1996-06-04)
Hydrogen peroxide oxidation of human erythrocytes induces a transfer of phospholipid from the membrane into the cytosol [Brunauer, L.S., Moxness, M.S., & Huestis, W.H. (1994) Biochemistry 33, 4527-4532]. The current study examines the mechanism of lipid reorganization in oxidized cells.
M R Hileman et al.
FEBS letters, 415(2), 145-154 (1997-11-14)
The neurotrophin receptor (NTR) and tumor necrosis factor receptor family of receptors regulate apoptotic cell death during development and in adult tissues [Beutler and van Huffel, Science 264 (1994) 667-668]. We have examined a fragment of p75NTR from the carboxyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service