Skip to Content
Merck
All Photos(1)

Key Documents

253510

Sigma-Aldrich

3-[5-(Dimethylamino)-1-naphthalenesulfonamido]-PROXYL

Synonym(s):

3-[5-(Dimethylamino)-1-naphthalenesulfonamido]-2,2,5,5-tetramethyl-1-pyrrolidinyloxy, free radical

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H29N3O3S
CAS Number:
Molecular Weight:
391.53
UNSPSC Code:
12352103

Features and Benefits

Spin label.

Legal Information

Licensed under U.S. Patent 3 489 522

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service