Skip to Content
Merck
All Photos(1)

Key Documents

252468

Sigma-Aldrich

Rhodamine 6G tetrafluoroborate

Dye content 99 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H30N2O3 · HBF4
CAS Number:
Molecular Weight:
530.36
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
Pricing and availability is not currently available.

composition

Dye content, 99%

mp

264-266 °C (lit.)

λmax

528 nm

SMILES string

F[B-](F)(F)F.CCNc1cc2OC3=CC(=[NH+]/CC)\C(C)=CC3=C(c2cc1C)c4ccccc4C(=O)OCC

Looking for similar products? Visit Product Comparison Guide

Application

Suitable as a laser dye

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service