Skip to Content
Merck
All Photos(1)

Documents

248428

Sigma-Aldrich

Magnesium methyl carbonate solution

0.5 M in methanol

Synonym(s):

MMC, Methoxymagnesium methyl carbonate, Methyl methoxymagnesium carbonate, Stile’s reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OMgOCO2CH3
CAS Number:
Molecular Weight:
130.38
Beilstein:
4342967
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

concentration

0.5 M in methanol

density

0.849 g/mL at 25 °C

SMILES string

CO[Mg]OC(=O)OC

InChI

1S/C2H4O3.CH3O.Mg/c1-5-2(3)4;1-2;/h1H3,(H,3,4);1H3;/q;-1;+2/p-1

InChI key

PNVKCUSCADAAMP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in Sure/Seal™ bottles

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service