Skip to Content
Merck
All Photos(1)

Key Documents

238953

Sigma-Aldrich

Drierite

without indicator, 20-40 mesh

Synonym(s):

Calcium sulfate, anhydrous

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12000000
Pricing and availability is not currently available.

particle size

20-40 mesh

Legal Information

Drierite is a trademark of W.A. Hammond Drierite Co., LTD

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enthalpies of vaporization of organic compounds.
Howard PB and Wadso I.
Acta Chemica Scandinavica, 24, 145-149 (1970)
The analysis of carbon monoxide in urban air at the ppm level, and the normal carbon monoxide value.
L Dubois et al.
Journal of the Air Pollution Control Association, 16(3), 135-139 (1966-03-01)
GAS CHROMATOGRAPHIC DETERMINATION OF LOWER ALCOHOLS IN BIOLOGIC SAMPLES.
H LYONS et al.
Clinical chemistry, 10, 429-432 (1964-05-01)
Respirometry: anhydrous drierite equilibrates with carbon dioxide and increases washout times.
White CR, et al.
Physiological and Biochemical Zoology : Pbz, 79(5), 977-988 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service