Skip to Content
Merck
All Photos(1)

Documents

238430

Sigma-Aldrich

1-Bromo-2-propanol

technical grade, 70%, contains MgO as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2Br
CAS Number:
Molecular Weight:
138.99
Beilstein:
1071195
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

70%

form

liquid

contains

MgO as stabilizer

refractive index

n20/D 1.480 (lit.)

bp

145-148 °C (lit.)

density

1.53 g/mL at 25 °C (lit.)

SMILES string

CC(O)CBr

InChI

1S/C3H7BrO/c1-3(5)2-4/h3,5H,2H2,1H3

InChI key

WEGOLYBUWCMMMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Conformational isomerism of 1-bromo-2-propanol has been investigated by NMR spectroscopy.

Other Notes

remainder 2-bromo-1-propanol

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F

Flash Point(C)

54 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karla M S Gonçalves et al.
The journal of physical chemistry. A, 117(42), 10980-10984 (2013-10-25)
The conformational isomerism of 1-chloro- (1) and 1-bromo-2-propanol (2) was theoretically and spectroscopically (NMR) analyzed. Conformers with the X-C-C-O (X = Cl and Br) fragment in the gauche orientation were found to be strongly prevalent both in the gas phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service